Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IRS1 Mouse Monoclonal Antibody [clone: 10I3]

Supplier: Boster
M00268-DYL488
76467-584EA 378.14 USD InStock
76467-584
Anti-IRS1 Mouse Monoclonal Antibody [clone: 10I3]
Antibodies
Mouse IgG monoclonal Anti-Human IRS1 Antibody DyLight®, 488 Conjugated, Flow Validated.

Type: Primary
Antigen: IRS1
Clonality: Monoclonal
Clone: 10I3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty