Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-KRT18 Mouse Monoclonal Antibody [clone: 7I6]

Supplier: Boster
M01357-4
76467-764EA 360.95 USD InStock
76467-764
Anti-KRT18 Mouse Monoclonal Antibody [clone: 7I6]
Antibodies
Mouse IgG monoclonal antibody for Cytokeratin 18 detection. Tested with WB, IHC-P, IHC-F, ICC/IF, IF, FCM in Human.

Type: Primary
Antigen: KRT18
Clonality: Monoclonal
Clone: 7I6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty