Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-RNH1 Mouse Monoclonal Antibody [clone: 4F3]

Supplier: Boster
M04147
76467-920EA 360.95 USD InStock
76467-920
Anti-RNH1 Mouse Monoclonal Antibody [clone: 4F3]
Antibodies
Mouse IgG monoclonal antibody for Ribonuclease Inhibitor/RNH1 detection. Tested with WB, IHC-P, ICC/IF, FCM in Human.

Type: Primary
Antigen: RNH1
Clonality: Monoclonal
Clone: 4F3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty