Would you like to log in again?
Western Blotting Mouse IgG DAB Chromogenic Reagent Kit (Yellow), Boster
Supplier: Boster
SA2020
76468-106EA 103.14 USD InStock
76468-106
Western Blotting Mouse IgG DAB Chromogenic Reagent Kit (Yellow), Boster
Blotting Kits
All-In-One kit with HRP conjugated secondary antibody and DAB substrate, optimized for Western Blotting.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty