Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VWR® Mega Star 1.6 General Purpose 1.6 L Benchtop Centrifuges

Supplier: VWR
VWR®
75009036 75883681 75883581 75009035
76519-280EA 9796.25 USD InStock
76519-280 76468-134 76468-132 76519-282
VWR® Mega Star 1.6 General Purpose 1.6 L Benchtop Centrifuges
Centrifuges Benchtop Centrifuges
Optimize projects with the Mega Star 1.6 general purpose centrifuge series. These versatile compact benchtop centrifuges provide maximum sample security and reproducibility in an intuitive, easy to use platform. Choose between a ventilated or refrigerated compact benchtop 1.6 L model, sold as an individual centrifuge or a package, to enhance productivity.

  • Space saving general purpose Mega Star 1.6 and 1.6R centrifuges
  • Equipped with a simple, intuitive keypad control interface in a compact footprint
  • Can run up to 16×50 ml conical tubes, 36×15 ml conical tubes or 10× standard microplates
  • Compatible with the Auto-Lock III Rotor system and ClickSeal biocontainment lids
  • Available as a tissue culture package that includes the TX-400 swinging bucket rotor, set of four 400 ml round buckets, set of four ClickSeal biocontainment lids, set of 4×50 ml conical and 15 ml conical tube adapters

Easy to Use: Intuitive controls simplify operation and provide detailed information on a wide range of operating parameters and processing status for optimal performance. Modern industrial design supports continuous improvement in rotor loading ergonomics, user safety and current needed certifications.

Auto-Lock™ III Rotor System: Secure locking system allows easy push-button installation and exchange of rotors. System saves time when switching between applications. Easy access to rotor chamber offers quick cleaning, a healthier working environment and longer unit life.

ClickSeal™ Bucket Sealing System: The ClickSeal Biocontainment solution - seals with a snap; eliminates screw caps and complicated clips. System has glove-friendly, one-handed open/close capability. System is certified by CAMR™ in Porton Down, UK.

Space-Saving Design: Compact dimensions optimize work area and ergonomic height and curved design ensure easy sample loading and unloading and easy cleaning.

Energy Efficient: System offers up to 40% energy consumption savings on industry standard protocols, such as standard blood separation or conical tubes processing.
Centrifuges have a 2 year warranty. Rotors have a different warranty period.

Ordering information: Centrifuge only option is supplied without accessories. Tissue Culture Packages includes TX400 rotor, set of four 400 ml buckets, set of four ClickSeal™ biocontainment lids, set of four 4×50 ml conical tube adapters and set of four 9×15 ml conical tube adapters.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty