Would you like to log in again?
Sterile Low Retention Filter Pipette Tips, Globe Scientific
Supplier: Globe Scientific
150935 150938 150900 150920 150903 150925 150910 150915
76468-282CS 271.89 USD InStock
76468-282 76468-280 76468-290 76468-278 76468-288 76468-276 76468-286 76468-284
Sterile Low Retention Filter Pipette Tips, Globe Scientific
Pipette Tips
Universal fit is compatible with most common brands of pipettors.
  • Manufactured from clear polypropylene (PP)
  • Polypropylene (PP) filters protect samples and pipettors from contamination
  • DNase-free, RNase-free and non-pyrogenic
  • Manufactured in a Class 100 000 cleanroom environment under ISO13485:2016 and ISO9001:2015 quality management systems
  • Sterilized by gamma irradiation SAL 10⁻⁶ (ISO 11137)
  • Racks are printed with lot number for quality traceability
Special processing renders these tips hydrophobic for exceptionally low sample retention. A universal fit design provides an airtight fit on most brands of pipettors.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty