Would you like to log in again?
PLATE 384 WELL BLACK W/FLAT BOTTOM CS100
Supplier: Corning
Plate, 384 well, Color: Black, with Clear Flat Bottom Polystyrene Poly-D-Lysine Coated Microplate, With Lid, with Custom Barcode, Aseptically Manufactured, Component Materials: Plate walls, Plate bottom, Lid, Poly-D-Lysine, Barcode Label-Virgin Polystyrene, meets USP, Class VI requirements
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
PLATE 384 WELL BLACK W/FLAT BOTTOM CS100 76468-866 9194BC Case of 100 Retrieving
Temperature sensitive Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty