Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

CellXpert® C170i Cell Culture CO₂ Incubators with O₂ Control

Supplier: Eppendorf
CellXpert®
6731011015 6731011015 6731011055 6731011815 6731011045 6731011815 6731011525 6731011515 6731011085 6731011355 6731011825 6731011025 6731011095 6731011825 6731011025 6731011095 6731011515 6731011345 6731011525 6731011355 6731011055 6731011085 6731011345 6731011045
76534-620EA 19444.61 USD InStock
76534-620 77321-638 CA77321-648 76468-918 CA77321-638 CA76534-616 76534-618 77321-646 CA77321-644 CA76534-614 77321-648 76534-616 CA77321-646 CA77321-650 77321-642 CA77321-640 76534-614 77321-644 CA76534-620 CA77321-642 CA76468-860 77321-650 77321-640 CA76534-618
CellXpert® C170i Cell Culture CO₂ Incubators with O₂ Control
Incubators CO₂ Incubators
Save costs and deliver results in your hypoxia experiments. The CellXpert C170i CO₂ incubator with O₂ control helps you save money (no expandable parts like HEPA filters, UV lamps, etc.). Keep contaminants away with the 180 °C High-Temp Disinfection. Grow your cells consistently with fast recovery, uniform temperature and no vibration from a fan.

  • Easy cleaning and disinfection: Seamless chamber, easy disassembling in 40 seconds, removable water tray
  • Certified, effective contamination protection: 180 °C High temperature disinfection (HTD) with protocol, no internal fan-associated HEPA filters necessary
  • Uniform temperature: Innovative heating concept with four temperature sensors at various locations verified at 27 locations inside the incubator (German DIN 12880:2007-05)
  • Fast recovery after door opening: Temperature and CO₂ in less than five minutes without regulation overshoot
  • Enhanced vibration and turbulence protection: No fan inside
  • Save money: No internal expendable parts (e.g. fan-associated HEPA filters), significantly reduced gas consumption, up to 25% more usable space by fanless design and innovative insulation concept
  • Stay flexible for the future: Upgrade your device in the laboratory later with various options
  • Full control and easy documentation: Advanced user interface, two front USB-ports, customized tasks and alarms, VisioNize® user management and connected lab

Today's cell culture requires more than technical solutions to keep operational costs under control, optimize cell growth conditions, and efficiently prevent contamination.

The CellXpert® CO₂ incubator family was designed with the needs of modern cell culture labs in mind - to reduce time-to-market or publication with minimal efforts and resources. They are the perfect partner for demanding applications like assay development or culture of stem cells, primary cells, or 3D organoids and tissues.

O₂ Sensor: ZrO₂ (zirconium dioxide)
Humidity delivery: Water tray
Copper package: Chamber, water tray and four shelves
CO₂ uniformity: ±0.1%

Certifications: GMP-/Cleanroom-related certificates and features: ISO class 4/Grade A cleanroom certified (ISO 14644-1), cytotoxicity test of materials: passed (ISO 10993-5:2009), production certificate, pdf event and parameter logging, HTD-effectiveness certificate and more.

Ordering information: Includes copper package, 4/8-segmented inner doors, door handle position, IQ/OQ GxP Service, VisioNize Lab Suite remote monitoring and more.

Order Now
 

ORDER

SPECIFICATIONS

ACCESSORIES

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty