Would you like to log in again?
Individual PCR Tubes, Globe Scientific
Supplier: Globe Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
PCR-02F PCR-05F PCR-05D PCR-02D
76468-958BX 58.4 USD InStock
76468-958 76468-964 76468-962 76468-960
Individual PCR Tubes, Globe Scientific
Tubes PCR Tubes
Individual PCR tubes are offered in 0.2 and 0.5 ml variants with either dome or flat caps. The individual PCR tubes are ideal for use in educational labs or when performing only a limited number of reactions.
  • Ultra-thin wall design (only 0.2 mm thick) for optimal temperature transfer
  • Easy open/close lids prevent sample loss
  • Attached flat or domed caps
Globe Scientific PCR plastics are engineered to bring a full range of solutions for routine end point PCR and qPCR (real-time) methods all the way to high-throughput automation demands.
Certified free of detectable Dnase, Rnase, Human DNA, PCR Inhibitors and tested Pyrogen-free.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty