Would you like to log in again?
Chix® Wet Wipes, Chicopee®
Supplier: Chicopee
Chix®
CHI8311 CHI8312
76469-060CT 121.82 USD InStock
76469-060 76469-046
Chix® Wet Wipes, Chicopee®
Wipes Disposable Wipes and Towels Presaturated Wipes
General-purpose towels are designed for use with water-based liquids.
  • Designed for use with water-based liquids
  • Nonwoven, low-lint material
  • Rinse-and-reuse design
Nonwoven, low-lint material allows use on most surfaces. Single-ply wipes are ideal for general-purpose cleaning applications.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty