Would you like to log in again?
Boardwalk® Hydrospun Wipers, Essendant
Supplier: Essendant
Boardwalk®
BWKP050JPW BWKP080JPR
76469-048RL 168.08 USD InStock
76469-048 76469-066
Boardwalk® Hydrospun Wipers, Essendant
Wipes Disposable Wipes and Towels Wipes for General Applications
Choose the strength and absorbency of durable hydrospun wipers for everyday tasks.
  • Highly durable and absorbent
  • Cloth-like feel
  • Resistant to many solvents
  • Ideal for use on surface prep, cleaning equipment, and tools
  • Great alternative to shop towels
Sturdy, with a cloth-like feel, these wipes are solvent-resistant for difficult work. Ideal for surface prep, cleaning equipment, and tools. Hydroentangled wood pulp and polypropylene material.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty