Would you like to log in again?
COVERALL RESPIRATOR FIT HOOD TAN L CS6
Supplier: CHEMLAND-LAKELAND INDUS
Coverall, respirator fit hood, double storm flap with hook and loop, elastic face and wrists, attached boots with boot flaps, Material: Spun bonded polypropylene, Color: Tan, Test Method: ASTM D3776, advanced chemical barrier with expanded chemical data and higher hold-out times, Size: large
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
COVERALL RESPIRATOR FIT HOOD TAN L CS6 76469-074 C4T165T-LG Case of 6 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty