Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
BioX Serological Pipettes, Tradewinds Direct
Supplier: TradeWinds (TWD)
BX07-5010 BX07-5025 BX07-5001 BX07-5050 BX07-5002 BX07-5005
76469-346CS 137.83 USD InStock
76469-346 76469-338 76469-348 76469-342 76469-344 76469-340
BioX Serological Pipettes, Tradewinds Direct
Pipettes Serological Pipettes
These high quality serological pipettes are DNase/RNase free and come individually wrapped.
  • Sterile
  • Individually wrapped, paper/plastic wrap
  • Non-pyrogenic
  • DNase/RNase free
  • Color coded
These sterile serological pipettes are not only DNase/RNase free and non pyrogenic, but they are also accurate to ±2% and color coded for your convenience.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty