Would you like to log in again?
NXSEQ SINGLE CELL RNA-SEQ BEAD 33MG
Supplier: Biosearch Technologies
NxSeq Single-cell RNA-seq Bead, High quality beads with barcodes, a primer binding site, UMI and a 3 oligo-dT capture sequence for single-cell mRNA sequencing applications using methods such as drop-seq, has tightly controlled bead size and produces consistent performance lot to lot, Size: 33mg
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
NXSEQ SINGLE CELL RNA-SEQ BEAD 33MG 76469-416 NX-SCB-1 Each Retrieving
Temperature sensitive Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty