Would you like to log in again?
Phenylalanine Ammonia-Lyase Activity Assay Kit (Fluorometric), BioVision
Supplier: BioVision
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
AB284571-100TEST
76469-554EA 809.86 USD InStock
76469-554
Phenylalanine Ammonia-Lyase Activity Assay Kit (Fluorometric), BioVision
Assays Enzyme Assays
To measure the activity of Phenylalanine Ammonia-Lyase in samples.
  • The assay is simple, sensitive and can detect as low as 3 µU (3 pmol substrate catalysis/minute) of PAL under assay conditions
  • A facile, rapid way to measure Phenylalanine ammonia-lyase activity in samples
Phenylalanine Ammonia-Lyase (PAL, EC 4.3.1.24) catalyzes the reversible non-oxidative deamination of L-phenylalanine to trans-cinnamic acid and ammonia without any external cofactors. PAL is expressed in plants, yeast and some fungi and is absent in bacteria and animals. Patients suffering from the genetic disorder, phenylketonuria is unable to metabolize phenylalanine that leads to irreversible cognitive impairment. Historically, phenylketonuria has been treated with a phenylalanine-restricted diet taken for the entirety of the patient’s life. Recently, a drug containing PEGylated PAL has been FDA-approved to treat phenylketonuria, limiting the need of a restricted diet.
BioVision's Phenylalanine Ammonia-Lyase Activity Assay Kit measures the activity of PAL via a two-step plate-based assay. The products of the PAL enzymatic reaction, generated in the first step, react with the developers resulting in the formation of a fluorescent compound, which is detected at Ex/Em = 410/470 nm. The kit can detect as low as 3 µUnits (3 pmol substrate catalysis per minute) of PAL under assay conditions.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty