Would you like to log in again?
illustra™ AutoScreen 96 Well Plates
Supplier: Cytiva
25900598 25900598
76469-804EA 1192.28 USD InStock
76469-804 CA76469-804
illustra™ AutoScreen 96 Well Plates
Nucleic Acid Reagents Sanger Sequencing Reagents
AutoScreen 96-well plate is used for purification of sequencing samples reactions prior to analysis on capillary array electrophoresis (CAE) sequencers such as MegaBACE 1000 or ABI Prism sequencing instruments.
  • Includes DNA Grade Sephadex G-50 rehydrated in double-distilled water as well as 0.05% Kathon™ CG/ICP Biocide added as a preservative
  • Preferred loading volume is 15 to 20 µl. The suggested range for loading volumes to achieve optimum purification is 10 to 20 µl
  • A plate lid may be used during this centrifugation step to improve consistency of volume recovery and reduce the likelihood of contamination during purification steps in sample sequencing
AutoScreen 96-Well Plates used with MegaBACE 1000 can show two to three times lower signal intensities compared with intensities from ethanol-precipitated reactions. This does not affect read lengths. AutoScreen 96 is also applicable to other size exclusion chromatography applications.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty