Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

General-Purpose pH Electrode, Glass Body, Cole Parmer

Supplier: Cole Parmer

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

05991-22 05991-22
76469-806EA 215.03 USD InStock
76469-806 CA76469-806
General-Purpose pH Electrode, Glass Body, Cole Parmer
Electrodes pH Electrodes
Faster response and increased stability across a pH range of 0 to 14.

  • More durable—dome bulb for better protection from breakage
  • Withstand higher temperatures and chemical attack
  • Get more stability in low-ionic strength solutions
  • Resistant to Na⁺ ion interferences
  • Compatible with most brands of pH meters

Glass body combination probes are able to withstand higher temperatures and chemical attack. Single-junction is ideal for clean samples.

Delivery information: Includes 1 m (3.3 ft.) cable with BNC connector.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty