Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
SP Bel-Art® Sterileware® Single Slot Powder Sampler, Bel-Art Products, a part of SP
Supplier: Bel-Art
Sterileware®
F37875-0620 F37875-0970
76469-978PK 285.66 USD InStock
76469-978 76469-980
SP Bel-Art® Sterileware® Single Slot Powder Sampler, Bel-Art Products, a part of SP
Samplers Solid Samplers
Ideal for taking a single, targeted 50 ml sample from free-flowing powders and granules.
  • After insertion, rotate tube to open then close the collection slot, and remove to retrieve the sample
  • Made from FDA and EU conforming materials; safe for food, drug and cosmetic use
  • High density polyethylene (HDPE) sampler heat sealed in individual polyethylene bags
  • Gamma irradiated sterile (SAL 10⁻⁶)
Lot stamped for accurate tracing; shipped with a Certificate of Processing for Sterilization.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty