Would you like to log in again?
SHARPS COLLECTOR SLIDE PHARMA 9GAL CS8
Supplier: BD
Sharps Collector, Pharmaceutical, Slide Top with Gasket or liquid-absorbing materials to prevent leakage, Visual, audible, tactile (VAT) perimeter locks to ensure that the lid is securely attached, Facilitate hands-free disposal, Dimension: 18.5 x 17.75 x 11.75, Size: 9gal
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
SHARPS COLLECTOR SLIDE PHARMA 9GAL CS8 76470-060 305329 Case of 8 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty