Would you like to log in again?
Whatman™ Uniflo™ Syringe Filters, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Whatman™
9931-3002 9924-3004 9911-2504 9918-2504 9927-3004 9920-1302 9909-2502 9931-3002 9916-1304 9913-2504 9920-2502 9912-1302 9920-2504 9912-2502 9932-3002 9915-2504 9913-2502 9931-3004 9921-1304 9918-2504 9910-1302 9910-1304 9929-3004 9911-2504 9924-3004 9910-1304 9909-1302 9910-2504 9925-3004 9921-2504 9914-2504 9909-2502 9914-2502 9930-3002 9930-3004 9913-2502 9929-3002 9928-3004 9932-3004 9920-1304 9924-3002 9909-2504 9912-2504 9911-2502 9910-2502 9923-3004 9916-1302 9925-3002 9927-3002 9911-1304 9921-2502 9923-3002 9932-3004 9910-2502 9931-3004 9914-2502 9910-1302 9923-3004 9915-2502 9911-1302 9928-3002 9926-3002 9911-1304 9929-3004 9912-1302 9921-2504 9920-2502 9911-1302 9932-3002 9920-2504 9910-2504 9909-1304 9909-1304 9926-3002 9913-2504 9921-1302 9920-1304 9912-2502 9927-3002 9926-3004 9923-3002 9921-1304 9925-3002 9924-3002 9917-2504 9912-1304 9914-2504 9911-2502 9921-2502 9909-1302 9927-3004 9925-3004 9930-3002 9909-2504 9926-3004 9930-3004 9928-3004 9912-2504 9928-3002 9917-2504 9929-3002 9921-1302 9920-1302 9912-1304
76470-070PK 1188.45 USD InStock
CA76470-080 76470-070 75849-374 CA75849-386 75849-372 CA76470-084 75849-378 76470-074 CA75849-382 CA76470-082 CA75849-384 76470-072 75849-376 CA76470-088 76470-078 CA76470-086 76470-076 75849-382 75849-380 75849-364 CA75849-374 76470-082 CA76470-090 75849-362 CA75849-376 76470-080 CA75849-370 75849-368 76470-086 CA76470-094 75849-366 CA75849-372 76470-084 CA76470-092 CA75849-378 CA76470-098 76470-088 CA76470-096 CA75849-380 75849-370 76470-092 76470-090 76470-096 76470-094 CA76211-004 CA76211-006 76423-278 CA76423-288 76423-274 CA76423-286 CA76423-284 76423-276 CA76423-282 76470-098 CA76423-280 76423-272 75849-386 75849-384 76211-004 76211-006 76211-008 CA76211-008 CA76423-278 CA76423-276 CA76423-274 CA76423-272 76211-010 76211-012 76211-014 76211-016 76423-290 76423-286 76423-288 76423-282 CA76423-290 76423-284 76423-280 CA75849-364 CA75849-366 CA75849-360 75849-356 CA75849-362 75849-358 CA76211-202 CA75849-368 CA76470-066 CA76470-064 75849-360 CA76470-102 CA76470-068 CA76470-100 76470-064 CA76470-072 CA76470-070 76211-202 CA75849-356 CA75849-358 76470-068 CA76470-076 76470-100 76470-066 CA76470-074 76470-102 CA76470-078
Whatman™ Uniflo™ Syringe Filters, Whatman products (Cytiva)
Filters Syringe Filters
Whatman™ Uniflo™ syringe filters are well suited for rapid, routine syringe filtration of small volume samples up to 100 ml in laboratory sample preparation.
  • Available in a variety of membrane choices and pore sizes with a PP overmold housing
  • Laser etched printing on the filter for easy identification
PVDF membranes are designed for high tensile strength, high solvent resistance, and low protein binding, making them suitable for biomedical filtration and HPLC sample preparation.
Nylon membranes are hydrophilic and are a good choice for aqueous and aqueous-organic samples. The membrane offers good chemical resistance to most common HPLC solvents, however it has limited resistance to acids, bases, halogenated hydrocarbons, aldehydes and strong oxidising agents. The most common application is HPLC sample filtration.
PTFE membrane filters are hydrophobic and will not allow water to pass without high pressures. Aqueous solutions may be filtered if the membrane is initially 'wetted' with alcohol or another appropriate solvent. PTFE membranes will stop aqueous aerosols in gas streams.
Packaging: Packaged in clean resealable, recyclable plastic boxes to optimize laboratory storage space.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty