Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Transport Tubes, Sterile, 10 ml, Globe Scientific

Supplier: Globe Scientific
6102S-R
76470-188CS 230.04 USD InStock
76470-188
Transport Tubes, Sterile, 10 ml, Globe Scientific
Tubes Test Tubes
Sterile 16×101 mm polypropylene (PP) transport tube with separate high-density polyethylene (HDPE) red screw cap. These leak-resistant tubes are tested to 70 Kpa and gamma sterilized. The self-standing tubes feature rounded bottoms with skirt.

  • Working volune: 10 to 13 ml
  • Brim capacity: 15 ml
  • Sterilized by irradiation
  • Self-standing
  • Non-graduated

The tubes are produced from polypropylene and are RNase, DNase and pyrogen free.

The included screw caps feature an interior recess that will clasp the back end of a swab for safety and convenience when performing nasal or nasopharyngeal sample collections.

These sterile tubes are bulk packed in bags of 500. The caps are also sterile and are packaged separately within the same case as the tubes.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty