Would you like to log in again?
Orum International Stainless Steel Satellite with HEPA Filter, contact plate, Hardy Diagnostics
Supplier: Hardy Diagnostics
BAS263K BAS262K
76470-204EA 3635.38 USD InStock
76470-204 76470-202
Orum International Stainless Steel Satellite with HEPA Filter, contact plate, Hardy Diagnostics
Samplers Air Samplers
Accessory option for ISOLATOR RABS (76377-520, 76377-522, 76377-524, 76377-526) instrument for facilities requiring HEPA-equipped microbial air samplers.
Ordering information: When using the HEPA satellite with the TRIO.BAS™ RABS ISOLATOR, all satellites must be of the same catalog number. The HEPA satellite format cannot be used concurrently with standard, or other, satellite configurations.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty