Would you like to log in again?
Dehydrated Culture Media, YPD (YEPD) Media, MP Biomedicals
Supplier: MP Biomedicals
114001022 114001232 114001222 114001032
76470-208EA 148.41 USD InStock
76470-208 76470-210 76470-214 76470-212
Dehydrated Culture Media, YPD (YEPD) Media, MP Biomedicals
Media for Microbiology Dehydrated Media for Microbiology
YPD Broth is used for routine growth of Saccharomyces cerevisiae for molecular biology studies.
  • Available in different packaging format - pre-measured capsules and bulk powder packing
  • YPD agar media contains agar in addition to the other ingredients
It is a nutritionally rich medium containing dextrose, peptone and yeast extract. Dextrose is the energy source. Yeast extract and peptone provide carbon, nitrogen, minerals, vitamins, trace ingredients and other essential growth nutrients which stimulate the growth of yeasts.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty