Would you like to log in again?
LIBERO CE PDF LOGGER FOR EXT PT100 PROBE
Supplier: ELPRO SERVICES, INC.
LIBERO CE, USB PDF Logger for external Pt100 probes (M8 connector). Temperature range -200 deg C.+400 deg C, multi use device, lifetime: 3 years, memory capacity: 75'500 values
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
LIBERO CE PDF LOGGER FOR EXT PT100 PROBE 76470-344 802279 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty