Would you like to log in again?
LIBERO ITS-COOL (2-8 C) PDF INDICATOR
Supplier: ELPRO SERVICES, INC.
LIBERO ITS - Cool (2-8 C), Multi-level PDF Indicator with internal probe. Measuring range -30 deg C.+70 deg C. Read-out possibility via Smartphone App to generates PDF with alarm statistics. Expiry date Management. Pre-configured with Standard Configuration Profile A001
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
LIBERO ITS-COOL (2-8 C) PDF INDICATOR 76470-346 802220 Box of 400 Retrieving
Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty