Would you like to log in again?
GLOVES CHEMICALRESISTANT 38612 SZ10 PK12
Supplier: Ansell Healthcare
Gloves, 38612, Polymer Chemical Resistant, Unsupported,Beaded Cuff, Smooth Grip, Black Butyl/Viton ( 4Mils Viton/8 Mils Butyl)11.8 Mil-12in, Size: 10
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
GLOVES CHEMICALRESISTANT 38612 SZ10 PK12 76470-368 38612100 Case of 36 (1/ Itm/Bx) Retrieving
Pack of 12 (1/Itm/Bx) Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty