Would you like to log in again?
TSV Lab Refrigerators, Freezers and Combination Units with Green Refrigerants, Thermo Scientific
Supplier: Thermo Scientific
TSV20FPSA TSV07CPSA TSV05CPSA TSV05RPSA TSV20RPSA TSV05RPAA TSV18CPSA
76449-110EA 4113.74 USD InStock
76449-110 76420-632 76470-398 76420-630 76470-396 76420-636 76420-634
TSV Lab Refrigerators, Freezers and Combination Units with Green Refrigerants, Thermo Scientific
Refrigerators
An economical SNAP-compliant solution for safe sample storage.
  • SNAP-compliant, environmentally friendly, hydrocarbon refrigerants
  • Factory tested prior to shipment
  • NRTL-listed
  • Adjustable temperature control (dial)
  • Easy-to-clean, durable finish
  • Manual and auto defrost options
Thermo Scientific™ TSV laboratory refrigerators, freezers and combination units with SNAP™-compliant, hydrocarbon refrigerant are particularly suited for routine laboratory applications with sizes ranging from small benchtop to full-height freestanding units. The new TSV models offer features that are not available in domestic models and meet the demanding requirements of the laboratory environment.
Certifications: cULus certified
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty