Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Benchtop Centrifuges, Megafuge ST1 and ST1R Plus and Packages, Thermo Scientific

Supplier: Thermo Scientific

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

75009271 75009271CAP 75017003CAP 75016002CAP 75016002 75016003 75017003 75017002 75009230 75016003CAP 75017002CAP 75016010 75009230CAP 75016010CAP 75016026USCAP 75016026US
76470-428EA 11459.83 USD InStock
CA76501-618 CA76501-604 CA76501-602 CA76501-612 CA76501-576 CA76501-620 CA76501-610 76470-428 CA76501-574 76470-438 76470-426 76470-414 76470-418 76470-442 76470-430 76470-454
Benchtop Centrifuges, Megafuge ST1 and ST1R Plus and Packages, Thermo Scientific
Centrifuges Benchtop Centrifuges
The Thermo Scientific™ Megafuge ST1/ST1R Plus benchtop centrifuges offer intuitive controls and versatility for power at every turn.

  • Auto-Lock rotor system
  • ClickSeal™ bucket sealing system
  • SMARTSpin™ technology
  • Space-saving design
  • Energy efficient

Secure locking system allows easy push-button installation and exchange of rotors. System saves time when switching between applications. Easy access to rotor chamber offers quick cleaning, a healthier working environment and longer unit life. Biocontainment solution seals with a snap; eliminates screw caps and complicated clips. System has glove-friendly, one-handed open/close capability. System is certified by CAMR™ in Porton Down, UK.

Advanced rotor management system maximizes acceleration, braking and residual load imbalance for each rotor and bucket option. Technology optimizes safety and improves separations.

Compact benchtop dimensions optimize working area, the ergonomic height and curved design with motorized lid latch ensure easy sample loading and unloading, easy cleaning and promotes user safety.

System offers up to 40% energy consumption savings on industry standard protocols, such as standard blood separation or conical tubes processing.

Certifications: IVD certified.

Ordering information: Centrifuge only, optional Cell Culture and Blood Tube packages are also available.
Cell Culture package includes TX400 rotor, set of four 400 ml buckets, set of four ClickSeal™ biocontainment lids, set of four 4×50 ml conical tube adapters, set of four 9×15 ml conical tube adapters.
Blood Tube package includes TX400 rotor, set of four 400ml buckets, set of four ClickSeal™ biocontainment lids, set of four 19×5/7 ml blood tube and 14×10 ml blood tube adapters.

Order Now
 

ORDER

SPECIFICATIONS

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty