Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VWR® Alliance Cleanroom Hood, Made in USA

Supplier: VWR

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

VWR®
350L 450M 450L 350M
76470-712CS 1397.52 USD InStock
76470-712 76470-706 76470-708 76470-710
VWR® Alliance Cleanroom Hood, Made in USA
Disposable Garments
VWR® Made in USA Cleanroom Hoods are an ideal option for single-use needs. Made with the specially formulated Alliance breathable microporous fabric.

  • Individually bagged
  • Available with bound or serged seams
  • White finish
  • Made with Alliance breathable microporous fabric
  • Made in USA

A critical part of the VWR® Made in USA PPE line, these cleanroom hoods provide a comfortable fit while meeting the standards of many controlled environments. Intended for Class 100 or higher enviroments.

The Alliance fabric is designed to lower particulate levels by providing an extremely smooth outer fabric surface that lowers particle cling and build-up in cleanroom applications. Alliance products have a unique anti-static system that assures static dissipation in sensitive cleanroom applications, while providing a liquid-proof barrier that ensures protection from hazardous substances used in Biotech and Pharmaceutical cleanrooms.

Packaging: 100 per case, packed in poly-lined bag within brown corrugated box.

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty