Would you like to log in again?
Monarch® Bead Retainers, New England Biolabs
Supplier: New England Biolabs
Monarch®
T3004L T3004L
76470-764EA 39 USD InStock
CA76470-764 76470-764
Monarch® Bead Retainers, New England Biolabs
Beads Glass Beads
Plastic parts for retaining the Monarch DNA capture beads during the Monarch HMW DNA extraction workflow. These pieces resemble spin columns without any membrane inserted and facilitate various steps of the extraction workflow. They mate to the Monarch collection tube II.
  • Supplied as a component of the Monarch HMW DNA extraction kits
  • Facilitates Monarch HMW DNA extraction workflow by containing the beads during wash and elution
  • Plastic part resembling a spin column without a membrane
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty