Would you like to log in again?
Monarch® RBC Lysis Buffer, New England Biolabs
Supplier: New England Biolabs
Monarch®
T3051L T3051L
76470-768EA 86 USD InStock
CA76470-768 76470-768
Monarch® RBC Lysis Buffer, New England Biolabs
Buffers Extraction Buffers
The Monarch® RBC lysis buffer is specifically designed to lyse red blood cells (erythrocytes) while leaving leukocytes intact. The separate RBC lysis step enables removal of hemoglobin to increase the purity of the blood prep. The RBC Lysis Buffer also removes nucleases to enable the subsequent purification of HMW DNA into the megabase (Mb) range in size.
  • A component of the Monarch HMW DNA extraction kit for cells and blood
Store this buffer at 4 °C.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty