Would you like to log in again?
Monarch® Protein Separation Solution, New England Biolabs
Supplier: New England Biolabs
Monarch®
T3062L T3062L
76470-774EA 74 USD InStock
CA76470-774 76470-774
Monarch® Protein Separation Solution, New England Biolabs
Buffers Extraction Buffers
The Monarch® Protein separation solution, supplied as a component of the Monarch HMW DNA extraction kit for tissue, is a salt-based solution that is optimized to ensure proteins in tissue and bacteria samples are easily removed during the phase separation of the Monarch HMW DNA extraction workflow.
  • Facilitates the separation of proteins during HMW DNA extraction from tissues
  • Salt-based solution for protein precipitation
Delivery information: Supplied in 2 bottles of 18 ml.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty