Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
Miltex® Needle Holders, Integra
Supplier: Integra LifeSciences
Miltex®
8-40TC 8-8TC ST8-44 ST8-50 8-44TC 8-46TC 8-42TC 8-4TC ST8-6
76470-900EA 250.14 USD InStock
76470-900 76470-898 76470-948 76470-904 76470-902 76470-908 76470-906 76470-952 76470-950
Miltex® Needle Holders, Integra
Forceps
A close examination of Miltex needle holders, either standard or tungsten carbide, will reveal meticulously crafted surgical tools that will perform precisely as intended.
Smoothly beveled box locks and jaw edges help to protect delicate suture material from snags or cutting. The jaw surface is milled to help ensure a secure grasp and to help prevent needle slippage or rotation.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty