Would you like to log in again?
Hughes ™ Eyewash Additive Solution, Justrite®
Supplier: Justrite
Hughes ™
ADDR4PKUS ADDR4PKCAN
76471-008PK 69.12 USD InStock
76471-008 CA76471-008
Hughes ™ Eyewash Additive Solution, Justrite®
First Aid Supplies Eyewash Stations and Eyewash Bottles
Eyewash additive preserves water in portable, self-contained eyewash stations to prevent bacterial growth.
  • Water preservative solution for use with self-contained eye/face wash stations
  • Each 8oz. bottle preserves 5 to 20 gal. of water for up to 120 days
  • Sealed bottle has a shelf life of 3 years
Eyewash additive treats water to help avoid additional damage to an already injured eye, while also extending the useful life of eyewash water. Labeled in accordance with USFDA.
Simply fill a clean eyewash station with potable water and add the additive solution. Treated water reduces the frequency of water changes, minimizing the maintenance required to stay compliant (ANSI Z358.1-2014, Section 5.5.3 recommends weekly visual checks of flushing fluid). A tamper-evident seal offers extra security.
For use with Hughes 10GFEW,16GFEW, 15GEW, 28GEW, and 38GEW and other portable or self-contained eyewashes.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty