Would you like to log in again?
Tester, R-C-L Meter, Excelta Corp®
Supplier: Excelta
TM-500 TM-500-BTW
76471-438EA 856.97 USD InStock
76471-438 76241-252
Tester, R-C-L Meter, Excelta Corp®
Resistance Testers
The TM-500 five star IntelliTweeze™ is a quick and easy way of testing and troubleshooting many types of components.
  • Easy way of testing and troubleshooting many types of components
It features built-in high precision LCR probe, 0.2 to 0.5% base measurement accuracy, automated component identification, automated test range selectioon, manual C,R,L,Z and ESR modes, adjustable test signal levels, diade polarity short testing, secondary D,Q, and ESR parameters, high visibility LED display and build-in Li-Ion battery.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty