Would you like to log in again?
TWEEZERS STRIPPING MAG WIRE
Supplier: EXCELTA CORPORATION
Tweezers, Magnet Wire Stripping, 36 AWG Magnet Wire, OAL: 4.5in 113mm110 angled, will strip magnet wire exactly to AWG gauge without nicking, made of carbon steel with flame hardened edges and coated with an Anti-Static (10 raised to 1 ⁰ ohms/sq) epoxy coating to prevent oxidation.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
TWEEZERS STRIPPING MAG WIRE 76471-500 15A-ST-PE-36M Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty