Would you like to log in again?
Pliers, Needle Nose, Excelta
Supplier: Excelta
2847LD 2847L 2647 2847L 2905 2847D-CR 2910 2905 2905D 2847SL 2847D 2910D 2647D 2647 2847 2647SD 2847 2847-CR
76471-612EA 40.4 USD InStock
76471-612 63041-810 76238-662 76471-614 63041-814 63041-726 63041-724 76238-670 76471-618 63041-812 94024-206 76471-586 75880-682 76471-610 76245-086 76246-954 76245-106 76238-668
Pliers, Needle Nose, Excelta
Pliers
The tapered jaws on these pliers allow better accessibility in tight places.
  • Needle nose for tight areas
  • Ergonomic handle, dual leaf springs
  • Stainless steel construction-corrosion resistant
  • Different sizes depending on application
Needle nose pliers have tapered noses for better accessibility in tight places. The noses aren't quite as strong as the 'Chain Nose' pliers due to the taper. Each of these is made of AISI 440 stainless steel to prevent oxidation. All include two leaf springs for more ergonomic control. They are offered in a small, medium or large head side, depending on the application. The 'D' specifies serrations on the tips for better control.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty