Would you like to log in again?
Miniature Wire Cutters, Excelta
Supplier: Excelta
9238SL 9231E 9231SL 9232SL 9238E 9232E
76471-634EA 106.82 USD InStock
76471-634 76471-636 76471-638 76471-640 76471-632 76471-642
Miniature Wire Cutters, Excelta
Cutters
These high-quality cutters are designed for cutting small soft wire up to 250,000 times without resharpening.
  • Three head shapes for various accessibility issues
  • Three types of cuts available
  • 'Set-Screw' stop prevents overtravel
  • Induction hardened blades for longevity
  • Ergonomic handles/dual leaf springs
These Lazer Line® side cutters are made of carbon tool steel with induction hardened cutting blades to RHC 63-65. The CNC machining allows the joint surfaces to be perfect to allow for long life. The dual leaf springs provide a smooth action. They feature an ergonomically molded antimicrobial grip (10¹⁰ ohms/sq surface resistivity) with dual leaf springs. The application for these tools is soft wire and after up to 200,000 cuts, these tools can be factory resharpened.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty