Would you like to log in again?
Sterilization Wraps for Steam, ETO or Gamma, Blue, Heavy Duty, Keystone Cleanroom Products
Supplier: Keystone Cleanroom Products
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
BHDSHEET2020 BHDSHEET3636 BHDSHEET3030 BHDSHEET0909 BHDSHEET5454 BHDSHEET5472 BHDSHEET1818 BHDSHEET1515 BHDSHEET4545 BHDSHEET4040 BHDSHEET2424
76471-688CS 624.05 USD InStock
76471-688 76471-690 76471-692 76471-680 76471-694 76471-682 76471-696 76471-684 76471-676 76471-698 76471-686
Sterilization Wraps for Steam, ETO or Gamma, Blue, Heavy Duty, Keystone Cleanroom Products
Sterilization Wraps
When secured with autoclave tape or elasticized bands, these blue BHD wrappers provide an effective microbial barrier from external sources of contamination.
  • Designed for steam sterilization; but also compatible with ETO or gamma irradiation
  • Treated to impart water and alcohol repellency
  • Wet formed, lightweight non-woven fabric comprised of natural wood pulp and polyester fibers bonded with a resin binder
  • Sufficient microbial barrier
  • Superior bacterial barrier properties compared to linen
  • Sterilization tapes adhere firmly and remove easily
  • Ideal choice for light trays, small basins and towels
  • Resists tearing and punctures
  • Does not contain any natural latex rubber
  • Processed and packaged in ISO certified cleanrooms
These versatile wraps offer fluid repellency and provide an excellent barrier against airborne bacteria. The soft, drapable non woven blend of cellulose and polyester is virtually lint-free and helps protect wrapped contents from contamination.
Delivery information: Certificates of Conformance supplied with each shipment.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty