Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Core 6D Carts, TrippNT

Supplier: TrippNT

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

53465 53473 53474 53470 53475 53467 53471 53463 53468 53464 53472 53466 53469
76471-766EA 1332.08 USD InStock
76471-766 76471-776 76471-768 76471-778 76471-780 76471-782 76471-760 76471-770 76471-784 76471-762 76471-772 76471-764 76471-774
Core 6D Carts, TrippNT
Carts
TrippNT's Core 6D offer 6 total locking drawers for flexible organization and storage.

  • 2 locking and 2 swivel 4" twin wheel casters, 6 full-extension total locking drawers, 6 side shelves (4 shelves, 2 dispensing shelves), tool/chart holder
  • Organizes, stores, and secures supplies
  • 100% total replacement lifetime guarantee/made in USA

It offers a tool holder for tablets, charts, and small utensils. Its built-in handles are ergonomic and easy to grab. It is constructed from a corrosion-resistant non-metal polymer (HDPE) with robust casters capable of easily navigating elevator gaps and uneven surfaces.

Ordering information: Available as fully assembled,13 color variants.

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty