Would you like to log in again?
Environmental Express® Oakton® 250 and 260 Series Handheld Meters, Cole-Parmer
Supplier: Cole Parmer
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Oakton®
35660-36 35660-48 35660-50 35660-30 35660-40 35660-32 35660-42 35660-38 35660-44 35660-46 35660-26 35660-28 35660-34 35660-52
76471-942EA 412.99 USD InStock
76471-942 76471-952 76471-944 76471-954 76471-946 76471-956 76471-934 76471-948 76471-958 76471-936 76471-960 76471-940 76471-950 76471-938
Environmental Express® Oakton® 250 and 260 Series Handheld Meters, Cole-Parmer
Electrochemical Meters pH and Multi-Parameter Meters
Reduce maintenance time with self-diagnostic function.
  • Rugged, waterproof, and dustproof with an IP 67 rating for use in any environment
  • Nonslip ergonomic design for comfortable handling
  • Easily view measurements with large monochrome LCD with white LED backlighting
  • Download and analyze data with USB output and phono jack printer connections
  • Take it with you—500 hours of battery life
Get easy-to-read and accurate readings with auto-hold, auto-stable, and real-time measurements with stability indicators. The Environmental Express® Oakton® 200 series of water quality meters features an electrode status indicator, which shows the condition of the electrode after each calibration. Easily view electrode condition, reading stability, measured parameter, and temperature in one screen. Don't worry about accuracy in fluctuating temperature—ATC ensures measurement accuracy. The simple user interface features adjustable auto shut-off time ranging from 1 to 30 minutes. Self-diagnosing meters display measurement and meter issues for fast fault finding and troubleshooting.
Easily take measurements and conveniently store meter and electrodes with built-in foldable stand on the back of the meter. Two grooves on the side of the meter serve as electrode holders, to hold up to 2 electrodes.
Choose from two series—the 250 series feature 500 data points while the 260 series features 1000 data points, a real-time clock, and USB communication.
Accessories information: For your convenience, cables, electrodes, and replacement parts are available for your Oakton meters.
Delivery information: Kits include meter, electrode, calibration solutions, and 2 AA batteries. Meter-only models include meter and 2 AA batteries.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty