Would you like to log in again?
VWR® Sterile Standard Square Weighing Boats, White
Supplier: VWR
VWR®
120710V 120880V
76472-584CS 614.51 USD InStock
76472-584 76540-490
VWR® Sterile Standard Square Weighing Boats, White
Boats Weighing Boats
These square weighing boats are flexible and feature smooth surfaces to provide accurate pour-outs with minimal sample loss.
  • Electron beam sterilized to reduce the risk of cross-contamination and compromised samples
  • Made from 100% virgin anti-static polystyrene that is waterproof and reduces the chance of materials sticking to the units
  • Disposable
  • Can be used with microwave moisture analyzers
  • Rounded corners simplify transfer and allow flexing for controlled pouring of liquid, powdered, or granular samples
  • Boats are shallow with a wide, flat bottom to resist tipping
  • FDA/food-grade compliant
Ideal for applications where there is concern of cross-contamination and ‘clean’ is a necessity in life science research, food, bio-pharmaceutical, environmental & molecular research environments.
Caution: Expires 3 years from the date of sterilization.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty