Would you like to log in again?
Valor® 1000 Compact Bench Scales, Food-Safe, Ohaus
Supplier: Ohaus
Valor®
30539393 30539390 30539392
76472-618EA 279.6 USD InStock
76472-618 76472-624 76472-622
Valor® 1000 Compact Bench Scales, Food-Safe, Ohaus
Balances and Scales
The Valor® 1000 models are reliable food safe scales, ideal for food preparation tasks in dry environments. The units are NSF-certified and the USDA AMS-accepted design supports HACCP systems.
  • Stainless steel platform
  • ABS housing incorporates convenient carry handles
  • Easy-to-operate via two-button operation
  • Large backlit LCD display
  • Portable with up to 1500 hour battery life (uses D-cell alkaline battery or power adapter)
With Legal for Trade certifications and a food-safe design that is NSF and HACCP certified, the Valor® 1000 series is a reliable choice for general food weighing. Easy-to-operate Valor® 1000 scales have external digital calibration software, an integrated level bubble and adjustable feet, selectable weighing units (gram; kilogram; pound; ounce; pound:ounce).
Intuitive two-button operation allows quick weight checking to simplify repetitive operations and increase operator efficiency. The energy-efficient design operates on batteries for up to one year or up to 1500 hours.
Certifications: Product Safety: IEC/EN 61010-1; UL certified power supply
Electromagnetic Compatibility: FCC Part 15 Class B; Canada ICES-003 Class B
Delivery information: Supplied with an AC adapter, requires 4 D (LR20) batteries which are not included). Optional stainless steel pans and plastic in-use covers are available for extra protection.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty