Would you like to log in again?
FRONTIER™ 5000 Series Micro Centrifuge, Ohaus
Supplier: Ohaus
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
FRONTIER™ 5000
30370692
76472-634EA 2616.72 USD InStock
76472-634
FRONTIER™ 5000 Series Micro Centrifuge, Ohaus
Centrifuges Benchtop Centrifuges
Dependable benchtop micro centrifuges, ideal for high-speed laboratory applications.
  • Running time: 10 sec to 99 hr 99 min 99 sec or continuous
Select from a variety of rotors designed for a range of laboratory applications. The FC5513 offers standard 24×1.5/2 ml microtube rotors with a biocontainment option. This centrifuge can also support spin columns and hematocrit capillary tubes when used with additional rotor options.
Modern touch-wheel control on a sleek, splash-proof interface - designed for ease of operation, the FC5513 features convenient control on a protective splash-proof interface, which allows for responsive operation - even when wearing gloves.
The compact design saves valuable benchtop space - enclosing all its functionalities within a small footprint, the FC5513 saves valuable space on the laboratory bench.
Certifications: Product Safety: IEC/EN 61010-1; IEC/EN 61010-2-020, Electromagnetic Compatibility: IEC/EN 61326-1 Class B, Basic environments; FCC Part 15 Class B, Compliance Mark: CE
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty