Would you like to log in again?
CHILLER RECIRCULATING 900W NON-STANDARD
Supplier: Thermo Fisher Scientific
Chiller, recirculating, non-standard voltage; Up to 900 watts (60Hz) 750 watts (50Hz) Air Cooled, Basic Controller, T1 Pump option are now available in low noise models. Quiet performance (<58 dBA) promotes a safe and reduced stress environment in your lab. 5C to 40C Temperature Range
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CHILLER RECIRCULATING 900W NON-STANDARD 76472-738 1013C10560210001 Each Retrieving
Temperature sensitive Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty