Would you like to log in again?
SWAB DUAL TIP HANDLE 1POINTED 1 FLAT BG1
Supplier: Texwipe
Swab, precision, dual-tipped, with one end pointed and the other flat, Head Width: 0.20 mm, Head Thickness: 0.20 mm, Head Length: 19.05mm, Handle Width: 2.50mm, Handle Thickness: 2.50mm, Handle Length: 114.30mm, Total Swab Length: 152.40mm
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
SWAB DUAL TIP HANDLE 1POINTED 1 FLAT BG1 76472-818 TX735 Bag of 1 Retrieving
Case of 20 Retrieving
Temperature sensitive Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty