Would you like to log in again?
SWAB PES STERILE KNIT LONG HANDLE CS1
Supplier: Texwipe
Swab, Sterile, Knit Polyester, Long handle, for cleaning broad surfaces and flat areas, Head Width: 7mm, Head Thickness: 2.8mm, Head Length: 16.8mm, Handle Width: 3.2mm, Handle Thickness: 3.2mm, Handle Length: 145.5mm, Total Swab Length: 162.3mm
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
SWAB PES STERILE KNIT LONG HANDLE CS1 76472-822 STX761 Case of 1 Retrieving
Temperature sensitive Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty