Would you like to log in again?
Revolve™ Sustainable Sterile Pre-Wetted Cleanroom Wipes
Supplier: Texwipe
Revolve™
STX1704P STX1709P STX1712P
76472-850BG 145.8 USD InStock
76472-850 TWSTX1709P 76472-846
Revolve™ Sustainable Sterile Pre-Wetted Cleanroom Wipes
Wipes Cleanroom Wipes Presaturated Cleanroom Wipes
Revolve™ sustainable wipes are made for the cleanroom industry and are composed of 100% upcycled polyester material with a sealed edge, processed on Texwipe's fully automated manufacturing system. Revolve™ wipes contribute to sustainability goals, reducing the amount of post-consumer water bottles going into landfills and reducing the level of carbon emissions.
  • Wipe and clean surfaces, equipment and parts
  • Apply and remove lubricants, adhesives, residues and other solutions including disinfectants
  • Clean with solvents such as isopropyl alcohol (IPA), ethanol, acetone and degreasers
Revolve™ processing provides low levels of ions, NVRs (non volatile residues), particles, and fibres for use in critical cleaning applications and environments. They are designed for high sorption capacity, which is ideal for spill control, cleaning, and solution application.
Revolve™ pre-wetted products are pre-wet with 0.2 µm filtered USP-grade 70% IPA/30% DIW. Pre-wetted wipes are packed in easy to use, recloseable slider bags, reducing solution evaporation that preserves the consistent wipes wetness level.
Certifications: Meet USP and USP wipe requirements. Sterile wipes are compatible for use in GMP grade A.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty