Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Rapid Response™ Multi-Drug Test Panels, BTNX

Supplier: BTNX
Rapid Response™
D12.8-1P29-25 D12.1-1P29-25 D3.4-1P29-25 D2.1-1P29-25 D12.2-1P29-25 D10.1-1P29-25 D5.1-1P29-25 D6.1-1P29-25
76473-440KT 46.18 USD InStock
76473-440 76473-450 76473-438 76473-404 76473-434 76473-422 76473-458 76473-424
Rapid Response™ Multi-Drug Test Panels, BTNX
Drug Tests
The Rapid Response® Multi-drug test panel is a rapid, one-step, qualitative test for the simultaneous detection of multiple drugs and metabolites in human urine.

  • Sample: Urine
  • Format: Panel
  • Time to result: 5 minutes
  • Storage Condition: 2 to 30 °C (36 to 86 °F)
  • Test principle: Lateral flow immunoassay

Multi-Drug Test Panels can include COC (300), AMP (1000), MET (1000), THC (50), MTD (300), OPI (2000), PCP (25), BAR (300), BZO (300), TCA (1000), MDMA (500), MOP (300), OXY (100), PPX (300), BUP (10), FYL (10).

Delivery information: Includes 25 test panels, SVT, adulterant color chart (if applicable) and product insert.

Caution: This test is intended for in vitro diagnostic use only.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty