Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Rapid Response™ Multi-Drug One Step Cups, BTNX

Supplier: BTNX
Rapid Response™
D12.26-1VA D12.5-1V D5.1-1V D10.14-1V D12.11-1VA
76473-430KT 131.54 USD InStock
76473-430 76473-452 76473-420 76473-426 76473-406
Rapid Response™ Multi-Drug One Step Cups, BTNX
Drug Tests
The Rapid Response® Multi-drug urine test cup is a rapid, qualitative test for the simultaneous detection of multiple drugs and metabolites in human urine at a specified cut off level.

  • Sample: Urine
  • Format: Cup
  • Time to result: 5 minutes
  • Storage Condition: 4 to 30 °C (39 to 86 °F)
  • Test principle: Lateral flow immunoassay

Interpretation of test results are: positive (one line), negative (two lines), invalid (no lines or no Control line); or the comparison of reaction pad color to the corresponding color blocks for the adulteration strips.

Multi-Drug Urine Test Cups can include COC (300), AMP (1000), MET (1000), THC (50), MTD (300), OPI (2000), OXY (100), PCP (25), BAR (300), BZO (300), MTD (300), MDMA (500), MOP (300), BUP (10), EDDP (100), ETG (500), FYL (10).

Adulteration strip combinations: Cre/pH/Oxi, Cre/SG/pH.

Delivery information: Includes test cups, product insert.

Caution: This test is intended for in vitro diagnostic use only.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty